missing translation for 'onlineSavingsMsg'
Learn More

basal cell adhesion molecule (Lutheran blood group), Mouse, Polyclonal Antibody, Abnova™

Product Code. 16110805
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16110805 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16110805 Supplier Abnova Supplier No. H00004059A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant BCAM.

Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: EVRLSVPPLVEVMRGKSVILDCTPTGTHDHYMLEWFLTDRSGARPRLASAEMQGSELQVTMHDTRGRSPPYQLDSQGRLVLAEAQVGDERDYVCVVRAGA

Specifications

Antigen basal cell adhesion molecule (Lutheran blood group)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant BCAM.
Formulation 50% glycerol
Gene BCAM
Gene Accession No. NM_005581
Gene Alias AU/CD239/LU/MSK19
Gene Symbols BCAM
Host Species Mouse
Immunogen BCAM (NP_005572, 32 a.a. ∼ 131 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4059
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.