missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519973
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is K562 cell lysate. PA5-19973 can be used with blocking peptide PEP-0098.
BCMA Polyclonal antibody specifically detects BCMA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BCMA | |
Polyclonal | |
Unconjugated | |
Tnfrsf17 | |
B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
608 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q02223 | |
Tnfrsf17 | |
A synthetic peptide mapping at the carboxy terminus of human BCMA. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |