missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Mouse anti-Human, Clone: T218, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA126523
This item is not returnable.
View return policy
Description
MA1-26523 detects Lewis B from human samples. MA1-26523 has been successfully used in immunohistochemistry (paraffin) procedures. The MA1-26523 immunogen is SK-CO-10 human colon cancer cell line.
Specifications
| MUC5AC | |
| Monoclonal | |
| Unconjugated | |
| P98088 | |
| Mouse | |
| IgM | |
| RUO | |
| 4586 | |
| 4° C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| T218 | |
| MUC5AC | |
| MUC5AC | |
| SK-CO-10 human colon cancer cell line. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction