missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLVRB, Mouse, Clone: 2F4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BLVRB.
Brand: Abnova H00000645-M09.100ug
This item is not returnable.
View return policy
Description
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).[supplied by OMIM
Sequence: VACTSAFLLWDPTKVPPRLQAVTDDHIRMHKVLRESGLKYVAVMPPHIGDQPLTGAYTVTLDGRGPSRVISKHDLGHFMLRCLTTDEYDGHSTYPSHQYQSpecifications
| BLVRB | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000713 | |
| BLVRB | |
| BLVRB (NP_000704, 107 a.a. ∼ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F4 | |
| Mouse monoclonal antibody raised against a partial recombinant BLVRB. | |
| BLVRB | |
| BVRB/FLR/MGC117413/SDR43U1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 645 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction