missing translation for 'onlineSavingsMsg'
Learn More

BMI-1 Mouse anti-Human, Alexa Fluor 488, Clone: P51-311, BD

Product Code. 15852099
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15852099 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15852099 Supplier BD Biosciences Supplier No. 562636

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The P51-311 monoclonal antibody binds to human BMI-1 (B lymphoma Mo-MLV insertion region 1 homolog). BMI1 is a c-myc cooperating oncogene that encodes an ∽45 kDa protein that is a member of the Polycomb Group (PcG) of proteins. PcG proteins are essential for the maintenance, but not initiation, of the transcriptionally repressed state of certain developmental genes. PcG proteins are a structurally diverse group of proteins with conserved functions from fly to human cells. PcG proteins form complexes and regulate the expression of genes involved in cell cycle, DNA repair and differentiation that are crucial for maintaining the self renewal of normal and cancer stem cells. Specifically, BMI-1 is a core component of PRC1 (polycomb repressive complex 1). BMI-1, via the up-regulation of hTERT and independent of c-myc, can immortalize mammary epithelial cells. BMI-1 has also been shown to repress the INK4A locus that controls the tumor suppressors p16 and p19ARF (mouse homologue of p14ARF) in mouse models. BMI-1 plays a role in maintaining the self-renewal capacities of stem cells including hematopoietic, intestinal, retinal and neural stem cells. During antibody development, the purified P51-311 monoclonal antibody was found to detect BMI-1 by Western blot analysis of cellular lysates and by indirect immunofluorescent staining and flow cytometric analysis of fixed and permeabilized cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BMI-1
Applications Flow Cytometry
Classification Monoclonal
Clone P51-311
Conjugate Alexa Fluor 488
Description BMI1; PCGF4; polycomb group ring finger 4; RING finger protein 51; RNF51
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BMI-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.