Learn More
BSCL2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BSCL2.
Brand: Abnova H00026580-A01.50uL
Description
This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: WGGIWPRHRFSLQVNIRKRDNSRKEVQRRISAHQPGPEGQEESTPQSDVTEDGESPEDPSGTEGQLSEEEKPDQQPLSGEEELEPEASDGSGSWEDAALSpecifications
| BSCL2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant BSCL2. | |
| BSCL2 | |
| GNG3LG/HMN5/MGC4694/SPG17 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_032667 | |
| BSCL2 | |
| BSCL2 (NP_116056, 259 a.a. ∼ 357 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 26580 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.