Learn More
BZRP, Mouse, Clone: 3D8-B2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BZRP.
Brand: Abnova H00000706-M01.100ug
Description
Present mainly in the mitochondrial compartment of peripheral tissues, the protein encoded by this gene interacts with some benzodiazepines and has different affinities than its endogenous counterpart. The protein is a key factor in the flow of cholesterol into mitochondria to permit the initiation of steroid hormone synthesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MAPPWVPAMGFTLAPSLGCFVGSRFVHGEGLRWYAGLQKPSWHPPHWVLGPVWGTLYSAMGYGSYLVWKELGGFTEKAVVPLGLYTGQLALNWAWPPIFFGARQMGWALVDLLLVSGAAAATTVAWYQVSPLAARLLYPYLAWLAFATTLNYCVWRDNHGWHGGRRLPESpecifications
| BZRP | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001110 | |
| TSPO | |
| BZRP (AAH01110.1, 1 a.a. ∼ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3D8-B2 | |
| Mouse monoclonal antibody raised against a full length recombinant BZRP. | |
| TSPO | |
| BZRP/DBI/IBP/MBR/PBR/PKBS/PTBR/mDRC/pk18 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 706 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.