missing translation for 'onlineSavingsMsg'
Learn More

Calcineurin Mouse, Unlabeled, Clone: 29, BD

Product Code. p-3855829
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815369 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815369 Supplier BD Biosciences Supplier No. 610259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calcineurin is a Ca2+/calmodulin-dependent protein phosphatase. The active enzyme is a heterodimer of a large calmodulin-binding catalytic subunit A (61kDa) and a smaller Ca2+ binding subunit B (19kDa). Regions corresponding to the calmodulin-binding site, an autoinhibitory domain, and a putative subunit B binding site have been identified within the large subunit A. The activity of calcineurin is sensitive to immunosuppressants such as cyclosporin A (CsA) and tacrolimus (FK506). The study of FK506-mediated inhibition of nitric oxide formation has revealed that nitric oxide synthase (NOS) is a calcineurin substrate. Calcineurin dephosphorylates NOS and enhances its catalytic activity. Therefore, Calcineurin is an essential mediator for efficient T cell antigen receptor (TCR)-mediated T cell activation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.