Learn More
CALCR, Mouse, Clone: 2F7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CALCR.
Brand: Abnova H00000799-M01.100ug
Description
This gene encodes a high affinity receptor for the peptide hormone calcitonin and belongs to a subfamily of seven transmembrane-spanning G protein-coupled receptors. The encoded protein is involved in maintaining calcium homeostasis and in regulating osteoclast-mediated bone resorption. Polymorphisms in this gene have been associated with variations in bone mineral density and onset of osteoporosis. Alternate splicing results in multiple transcript variants. [provided by RefSeq
Sequence: CNNEVQTTVKRQWAQFKIQWNQRWGRRPSNRSARAAAAAAEAGDIPIYICHQELRNEPANNQGEESAEIIPLNIIEQESSASpecifications
| CALCR | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001742 | |
| CALCR | |
| CALCR (NP_001733.1, 394 a.a. ∼ 474 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F7 | |
| Mouse monoclonal antibody raised against a partial recombinant CALCR. | |
| CALCR | |
| CRT/CTR/CTR1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 799 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.