Learn More
calpain 1, (mu/I) large subunit, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CAPN1.
Brand: Abnova H00000823-A01.50uL
Description
The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. [provided by RefSeq
Sequence: FNILWNRIRNYLSIFRKFDLDKSGSMSAYEMRMAIESAGFKLNKKLYELIITRYSEPDLAVDFDNFVCCLVRLETMFRFFKTLDTDLDGVVTFDLFKWLQLTMFASpecifications
| calpain 1, (mu/I) large subunit | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CAPN1. | |
| CAPN1 | |
| CANP/CANP1/CANPL1/muCANP/muCL | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC008751 | |
| CAPN1 | |
| CAPN1 (AAH08751, 610 a.a. ∼ 714 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 823 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.