Learn More
CCR6 Rat anti-Mouse, Alexa Fluor 647, Clone: 140706, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557976
Description
The 140706 monoclonal antibody reacts with mouse CD196 (CCR6), a CC chemokine receptor that is expressed on B lymphocytes and subsets of T and dendritic cells. The known ligand for CCR6 is CCL20/MIP-3α, and together they regulate the migration and recruitment of antigen-presenting and immunocompetent cells during inflammatory and immunological responses.
Specifications
| CCR6 | |
| Monoclonal | |
| 0.2mg/mL | |
| CCR6, BN-1, CKR-L3, DRY6, KY411, GPR-CY4, STRL22 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 140706 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CCR6-transfected Rat Cell Line | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.