Learn More
CD10 Mouse anti-Human, BB515, Clone: HI10a, BD
Mouse Monoclonal Antibody
€ 268.36
Specifications
| Antigen | CD10 |
|---|---|
| Clone | HI10a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
Description
The HI10a monoclonal antibody specifically binds to CD10 which is also known as Neutral endopeptidase (NEP), Enkephalinase, Atriopeptidase, and Neprilysin. CD10 is encoded by MME (membrane metallo-endopeptidase). CD10 is a 100 kDa type II transmembrane glycoprotein that has neutral endopeptidase activity and is otherwise known as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). CD10 is expressed on a wide variety of normal and neoplastic cell types. Normal cells expressing CD10 include granulocytes, bone marrow stromal cells, a subset of B-cell progenitors, germinal center B cells and fibroblasts. This cell surface metalloendopeptidase inactivates a number of signaling molecules and serves as a major regulator in the nervous, immune and other systems.
Specifications
| CD10 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| IgG1 κ | |
| Affinity Purified | |
| MME; CALLA; EPN; NEP; neprilysin; SFE; atriopeptidase; enkephalinase |
| HI10a | |
| Monoclonal | |
| Mouse | |
| Human | |
| Acute CALLA Leukemia Blast Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.