Learn More
CD10 Mouse anti-Human, Brilliant Violet 605, Clone: HI10a, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 562978
Product Code. 15842409
  • ONLINE EXCLUSIVE PRICE
    € 439.00
    € 438.70 / Each
    Save € 0.30
    0.07% Off
Description
The HI10a monoclonal antibody specifically binds to CD10 which is also known as Neutral endopeptidase (NEP), Enkephalinase, Atriopeptidase, and Neprilysin. CD10 is encoded by MME (membrane metallo-endopeptidase). CD10 is a 100 kDa type II transmembrane glycoprotein that has neutral endopeptidase activity and is otherwise known as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). CD10 is expressed on a wide variety of normal and neoplastic cell types. Normal cells expressing CD10 include granulocytes, bone marrow stromal cells, a subset of B-cell progenitors, germinal center B cells and fibroblasts. This cell surface metalloendopeptidase inactivates a number of signaling molecules and serves as a major regulator in the nervous, immune and other systems.
For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment. Fluorescent dye interactions may cause staining artifacts which may affect data interpretation. The BD Horizon Brilliant Stain Buffer was designed to minimize these interactions. More information can be found in the Technical Data Sheet of the BD Horizon Brilliant Stain Buffer (Cat. No. 563794).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD10
Monoclonal
Brilliant Violet 605
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Acute CALLA Leukemia Blast Cells
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
HI10a
MME; CALLA; EPN; NEP; neprilysin; SFE; atriopeptidase; enkephalinase
Mouse
Affinity Purified
RUO
Human
IgG1 κ