missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD106 Rat anti-Mouse, Unlabeled, Clone: 429 (MVCAM.A), BD
Rat Monoclonal Antibody
Brand: BD Biosciences 550547
This item is not returnable.
View return policy
Description
The 429 monoclonal antibody specifically binds to both the long (∼110 kDa) transmembrane-spanning form and the truncated (∼47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α; 4 β; 1 integrin) and LPAM-1 ( α; 4 β; 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.
Specifications
| CD106 | |
| Monoclonal | |
| 31.25μg/mL | |
| Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 429 (MVCAM.A) | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Mouse preadipose cell line PA6 | |
| 1 mL | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction