missing translation for 'onlineSavingsMsg'
Learn More

CD110 Mouse anti-Human, APC, Clone: 1.6.1, BD

Product Code. 15861719
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15861719 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861719 Supplier BD Biosciences Supplier No. 562199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 1.6.1 monoclonal antibody specifically binds to the human Thrombopoietin Receptor (TPO-R) that is also known as the Myeloproliferative leukemia protein (c-Mpl) or CD110. CD110 is a type I transmembrane glycoprotein and a member of the hematopoietin receptor family. It is expressed on hematopoietic stem cells, a subfraction of hematopoietic precursor cells, cells of the megakaryocytic lineage and platelets. CD110 serves as a receptor for thrombopoietin. Upon binding of thrombopoietin to CD110, megakaryocyte proliferation and differentiation is induced, platelets are produced and stem cells are protected from apoptosis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD110
Applications Flow Cytometry
Classification Monoclonal
Clone 1.6.1
Concentration 0.2mg/mL
Conjugate APC
Description C-MPL; MPL; MPLV; TPOR; Thrombopoietin receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.