missing translation for 'onlineSavingsMsg'
Learn More

CD11b Mouse anti-Rat, APC, Clone: WT.5, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 562102

Product Code. 15811649

  • ONLINE EXCLUSIVE PRICE
    € 240.00
    € 239.68 / Each
    Save € 0.32
    0.13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The WT.5 monoclonal antibody specifically binds to CD11b, the α subunit of Mac-1 found on neutrophils and myeloid cells, but not on lymphocytes. Mac-1 expression is rapidly upregulated on neutrophils after activation, in the same time period that CD62L (L-selectin) is shed from the cell surface. Mac-1 mediates adhesion to ICAM-1 (CD54) and C3bi. Neutrophil binding to ICAM-1 is blocked by mAb WT.5.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11b
Monoclonal
0.2mg/mL
Itgam; Integrin αM chain; Mac-1 α chain; Integrin beta 2 alpha subunit
Mouse
Affinity Purified
RUO
Rat
IgA κ
Flow Cytometry
WT.5
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
Rt neutrophils
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.