Learn More
CD11b/Mac-1 Mouse anti-Human, BUV395, Clone: ICRF44 (also known as 44), BD
Mouse Monoclonal Antibody
€ 210.79 - € 425.00
Specifications
| Antigen | CD11b |
|---|---|
| Clone | ICRF44 (also known as 44) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 395 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15803149
|
BD Biosciences
563840 |
25 Tests |
€ 217.00 € 210.79 / Each Save € 6.21 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15893139
|
BD Biosciences
563839 |
100 Tests |
€ 425.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β 2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.
The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Specifications
| CD11b | |
| Flow Cytometry | |
| Brilliant Ultraviolet 395 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Mac-1α, integrin αM subunit, CR3 α chain |
| ICRF44 (also known as 44) | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human monocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.