missing translation for 'onlineSavingsMsg'
Learn More

CD11c Hamster anti-Mouse, FITC, Clone: HL3, BD

Product Code. 15887398
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887398 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887398 Supplier BD Biosciences Supplier No. 553801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hamster Monoclonal Antibody

The HL3 monoclonal antibody specifically binds to the integrin αx chain of gp150, 95 (CD11c/CD18). CD11c is expressed on dendritic cells, CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and some NK cells. It is upregulated on IEL and lymph-node T cells following in vivo activation. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone HL3
Concentration 0.5mg/mL
Conjugate FITC
Description Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.