Learn More
CD11c Hamster anti-Mouse, PE, Clone: HL3, BD
Description
The HL3 antibody reacts with the integrin αx chain of gp150, 95 (CD11c/CD18) which is expressed on dendritic cells and CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and is upregulated on IEL and lymph-node T cells following in vivo activation. CD11c is also found on human NK cells. Although its expression on mouse NK cells is not published, we have detected CD11c on mouse splenic NK cells. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.
Host Species: Hamster
Clone: HL3
Isotype: IgG1, λ
Cross Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | CD11c |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HL3 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Description | Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4 |
| Formulation | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Host Species | Hamster |
| Immunogen | C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.