missing translation for 'onlineSavingsMsg'
Learn More

CD124 Rat anti-Mouse, NA/LE, Unlabeled, Clone: MIL4R-M1, BD

Product Code. 15856778
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15856778 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15856778 Supplier BD Biosciences Supplier No. 552288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The mIL4R-M1 monoclonal antibody specifically binds to CD124 which is also known as the α subunit of the mouse Interleukin-4 Receptor (IL-4Rα). The mouse IL-4Rα is a 140kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and nonhematopoietic cells and cell lines. The cell surface IL-4Rα chain binds IL-4 with high affinity and associates with either the common γ chain (IL-4Rα/γc; aka, type I IL-4R) or the IL-13 receptor alpha subunit (IL-4Rα/IL-13Rα; aka, type II IL-4R complex) to form two distinct types of signal-transducing IL-4R complexes. The type I IL-4 receptor complex specifically binds IL-4 whereas the type II IL-4R binds and transduces signals from either IL-4 or IL-13. The mIL4R-M1 antibody blocks IL-4 binding to cells and is reported to be a potent inhibitor of IL-4 biological activities. The mIL4R-M1 antibody also recognizes naturally-occurring, soluble truncated forms of IL-4Rα (sIL-4R) that result either from enzymatic cleavage of the cell surface extracellular IL-4Rα domain or from differential mRNAsplicing and secretion by cells. These sIL-4R retain their high-affinity ligand binding domain and appear to either enhance or inhibit IL-4-mediated functions depending on the relative local levels of IL-4 and sIL-4R.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD124
Applications Flow Cytometry
Classification Monoclonal
Clone MIL4R-M1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Il4r; IL-4Rα; Il4ra; IL-4RA; IL-4R-alpha; IL4-BP; IL-4 Receptor α chain
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen Mouse CTLL-19.4 T Cell Line
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.