missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD137 Mouse anti-Human, APC, Clone: 4B4-1, BD
Mouse Monoclonal Antibody
€ 84.12 - € 465.41
Specifications
| Antigen | CD137 |
|---|---|
| Clone | 4B4-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15871249
|
BD Biosciences
561702 |
25 Tests |
€ 88.20 € 84.12 / Each Save € 4.08 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15816428
|
BD Biosciences
550890 |
100 Tests |
€ 466.00 € 465.41 / Each Save € 0.59 0.13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 4B4-1 monoclonal antibody specifically binds to CD137 which is also known as 4-1BB, and ILA (induced by lymphocyte activation). CD137 is a type I transmembrane glycoprotein that belongs to the TNF/NGF receptor family. It is encoded by TNFRSF9 (tumor necrosis factor receptor superfamily, member 9). CD137 is expressed on activated T cells, B cells, monocytes, and follicular dendritic cells. CD137 plays roles in the costimulation, differentiation and survival of T cells and B cells.Specifications
| CD137 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| 4-1BB |
| 4B4-1 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Recombinant Human 4-1BB Fusion Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title