Learn More
CD14 Mouse anti-Human, Brilliant Violet 510, Clone: MφP9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563079
Description
The MφP9 monoclonal antibody specifically binds to human CD14, a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, this CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| CD14 | |
| Monoclonal | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Monocytes | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MφP9 | |
| LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.