Learn More
CD15 Mouse anti-Human, APC, Clone: HI98, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551376
Description
The HI98 monoclonal antibody specifically reacts with 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten, SSEA-1, CD15 or Lewis X. This structure is found on a variety of cell surface glycolipids and glycoproteins. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and to a varying degree on monocytes, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. This antibody is also suitable for staining formalin-fixed, paraffin-embedded tissue sections without pretreatment. Since the Abs are recognizing a carbohydrate epitope (3-fucosyl-N-acetyllactosamine) they also should work across species and not only for human.
Specifications
| CD15 | |
| Monoclonal | |
| APC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HI98 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Mononuclear Cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.