Learn More
CD15 Mouse anti-Human, V450, Clone: MMA, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 642917
Description
CD15, clone MMA, is derived from the hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the U-937 histiocytic cell line.
CD15 recognizes a human myelomonocytic antigen. The structure recognized by CD15 antibodies is lacto-N-fucopentose III.
Specifications
| CD15 | |
| Monoclonal | |
| 100μg/mL | |
| HEPES buffer with gelatin and 0.1% sodium azide. | |
| U-937 histiocytic cell line | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| MMA | |
| Violet Fluor 450 (V450) | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.