Learn More
CD160 Mouse anti-Human, PE, Clone: BY55, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562118
Description
The BY55 monoclonal antibody specifically binds to the human cell surface molecule, CD160 (also known as BY55). CD160 is a glycosyolphosphatidylinositol-anchored glycoprotein member of the immunoglobulin superfamily. It is expressed on cytotoxic αβ CD8+ T cells and natural killer (NK) cells, γδ T cells and intestinal intraepithelial T lymphocytes. CD160 can bind to classical and nonclassical MHC class I molecules. This interaction can costimulate the proliferation of activated human T lymphocytes including CD28-negative T lymphocytes.
Specifications
| CD160 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human YT2C2 NK Cell Line | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| BY55 | |
| BY55; BY55 antigen; NK1; NK28 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.