missing translation for 'onlineSavingsMsg'
Learn More

CD18 Rat anti-Mouse, NA/LE, Unlabeled, Clone: GAME-46, BD

Product Code. 15858038
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15858038 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15858038 Supplier BD Biosciences Supplier No. 555280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The GAME-46 antibody reacts with the common β2 chain of LFA-1 (CD11a/CD18, αLβ2 integrin), Mac-1 (CD11b/CD18, αMβ2 integrin), and gp150, 95 (CD11c/CD18, αXβ2 integrin). Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with each of the three integrin α chains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. The β2 integrins are important mediators of leukocyte-endothelium interactions. It has been reported that GAME-46 antibody blocks in vitro adhesion of LFA-1-expressing cells to the ligands ICAM-1, ICAM-2 and ICAM-3 and of Mac-1-expressing cells to ICAM-1, C3bi, and fibrinogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone GAME-46
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Integrin β2 chain
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen LFA-1 (CD11a/CD18) from mouse T-cell hybridoma TAM8C41
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.