missing translation for 'onlineSavingsMsg'
Learn More

CD183 Mouse anti-Human, Alexa Fluor 488, Clone: 1C6/CXCR3, BD

Product Code. 15851279
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15851279 25 μg Each
15829178 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15851279 Supplier BD Biosciences Supplier No. 561730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Clone 1C6/CXCR3 reacts with the human form of the 40 - 41kDa seven-transmembrane protein and member of the G protein-coupled receptor family. CXCR3 is expressed primarily on activated T cells that infiltrate inflammatory sites. It has also been detected on some circulating T cells, B cells, and NK cells. Reports show that some CXCR3 T cells also express CCR5 and are mostly CD45RO cells. Three ligands for CXCR3 have been identified. They are interferon-γ inducible 10kDa protein (IP-10), monokine induced by interferon-γ (MIg), and interferon-inducible T-cell γ chemoattractant (I-TAC). These chemokines are produced by a variety of cells upon stimulation by IFN γ and interact with CXCR3 to mediate T-cell chemotaxis. This reagent has been reported to be suitable for immunohistochemical staining of acetone-fixed, frozen sections and/or formalin-fixed, paraffin-embedded tissue sections with citrate pretreatment. Clone CXCR3 also cross reacts with a subset of peripheral blood lymphocytes of baboon, and both rhesus and cynomolgus macaque monkeys. The distribution of lymphocytes is similar to that observed with peripheral blood lymphocytes from normal human donors. CXCR3 has been clustered as CD183 in the VIIth HLDA workshop. This product is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 1C6/CXCR3
Isotype: IgG1, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD183
Applications Flow Cytometry
Classification Monoclonal
Clone 1C6/CXCR3
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Description CXCR3; C-X-C chemokine receptor type 3; GPR9; IP10R; MigR; CKRL2; CMKAR3
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CXCR3 Peptide
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.