Learn More
CD1A Mouse anti-Human, Brilliant Violet 510, Clone: HI149, BD
Mouse Monoclonal Antibody
€ 210.06 - € 462.00
Specifications
| Antigen | CD1a |
|---|---|
| Clone | HI149 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 510 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15802829
|
BD Biosciences
563482 |
25 Tests |
€ 216.00 € 210.06 / Each Save € 5.94 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15892819
|
BD Biosciences
563481 |
100 Tests |
€ 475.00 € 462.00 / Each Save € 13.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HI149 monoclonal antibody specifically binds to CD1a. CD1a is a type I transmembrane glycoprotein. The 49 kDa CD1a polypeptide is associated with β2-microglobulin. CD1a is expressed on cortical thymocytes, dendritic cells and Langerhans cells. CD1a has structural similarities to the MHC class I antigen, and plays a role in antigen presentation.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| CD1a | |
| Flow Cytometry | |
| Brilliant Violet 510 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HI149 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| CD1; T6/Leu-6; R4; T6; HTA-1; FCB6 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.