Learn More
CD1c Mouse anti-Human, Alexa Fluor 647, Clone: F10/21A3, BD
Mouse Monoclonal Antibody
€ 115.70 - € 255.70
Specifications
| Antigen | CD1c |
|---|---|
| Clone | F10/21A3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15804049
|
BD Biosciences
565049 |
25 Tests |
€ 116.00 € 115.70 / Each Save € 0.30 0.26% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15894039
|
BD Biosciences
565048 |
100 Tests |
€ 260.00 € 255.70 / Each Save € 4.30 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The F10/21A3 monoclonal antibody specifically binds to CD1c. The CD1 family of transmembrane glycoproteins are structurally related to the classical major histocompatibility complex (MHC) proteins. CD1c is a type I transmembrane glycoprotein that forms heterodimers with beta-2-microglobulin. CD1c presents lipids and glycolipids of self or microbial origin to T cells. CD1c is expressed by Langerhans cells, dendritic cells, monocytes, cortical thymocytes, T cells, and some B cells.
Specifications
| CD1c | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| IgG1 κ | |
| Affinity Purified | |
| CD1; R7; M241; BDCA1 |
| F10/21A3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| GM-CSF- and IL-4-activated Human Monocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.