Learn More
CD20 Mouse anti-Human, BB515, Clone: 2H7, BD
Mouse Monoclonal Antibody
€ 112.35 - € 268.94
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15833729
|
BD Biosciences
564569 |
25 Tests |
€ 113.00 € 112.35 / Each Save € 0.65 0.58% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15823729
|
BD Biosciences
564568 |
100 Tests |
€ 277.00 € 268.94 / Each Save € 8.06 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
| CD20 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| IgG2b κ | |
| Affinity Purified | |
| MS4A1; B1; Bp35; LEU-16; S7 |
| 2H7 | |
| Monoclonal | |
| Mouse | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Human 6.16c1.3 B cell line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.