Learn More
CD201 Rat anti-Human, PE, Clone: RCR-252, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557950
Description
The RCR-252 monoclonal antibody specifically binds to the endothelial cell protein C receptor (EPCR), which is also known as CD201. CD201 is a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. CD201 binds activated protein C, a serine protease that is involved in regulating blood coagulation. EPCR is reportedly expressed on several malignant cell lines. Malignant cell lines that express EPCR can show significant levels of protein C activation and this activation can be inhibited by anti-EPCR antibody
Specifications
| CD201 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| EPCR-positive RE-1 Cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| RCR-252 | |
| EPCR; Activated protein C receptor; APC receptor; PROCR; centrocyclin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.