Learn More
CD209a Rat anti-Mouse, APC, Clone: 5H10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 564928
Description
The 5H10 monoclonal antibody specifically binds to mouse CD209a, or CIRE, which is the most closely related homolog of human DC-SIGN. In the mouse, several homologs of human CD209 (DC-SIGN) have been described. CD209a is a 238-amino acid type-II transmembrane C-type lectin, with an extracellular carbohydrate recognition domain. It can bind to mannosylated ligands in a Ca++ dependent manner. CD209a is expressed by some conventional CD4+ and CD4-CD8- dendritic cells (DC) but not by CD8+ DC. It is also expressed on monocytes, splenic plasmacytoid pre-DC, macrophages, and resident peritoneal cells. CD209a expression is downregulated upon cellular activation. CD209a is thought to play a role in the internalization and presentation of antigens to initiate immune responses.
Specifications
| CD209a | |
| Monoclonal | |
| 0.2mg/mL | |
| CD209; Cire; DC-SIGN; Dcsign; DC-SIGN1; CDSIGN | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 5H10 | |
| APC | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| Peptide Derived from Extracellular Region of CD209a/CIRE | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.