Learn More
CD247 (pY142) Mouse anti-Human, Unlabeled, Clone: K25-407.69, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558402
Description
The T cell receptor (TCR), expressed by thymocytes and T lymphocytes, is a multi-component cell-surface complex responsible for recognizing antigen in the context of MHC molecules. The antigen-specific binding component of the TCR, Ti, is a heterodimer of the variable Ig-like subunits a and b or g and d . Ti is non-covalently associated with an invariant set of molecules referred to as the CD3 polypeptides, g, d, e, and ζ ) was named CD247 at the 7th Human Leukocyte Differentiation Antigens Workshop. CD3 appears early in thymocyte differentiation and remains expressed on all mature T lymphocytes. After antigen recognition by the TCR, CD3 z is the primary intracellular signal transducing subunit. It contains three ITAMs ( I mmunoreceptor T yrosine-based A ctivation M otifs), each of which contains a pair of tyrosine residues that are phosphorylated by Lck and Fyn and are required for signal propagation. The molecular weight of CD3 z is 16 kDa, and it is also observed as 32-kDa homodimers or as heterodimers with the g chain of Fc receptors. Upon phosphorylation, the CD3 z monomer undergoes an apparent shift in electrophoretic mobility up to 21 kDa.
The K25-407.69 monoclonal antibody recognizes the phosphorylated tyrosine 142 (pY142) in the third ITAM domain of human CD3 z (CD247) .
Specifications
| CD247 (pY142) | |
| Monoclonal | |
| 0.5mg/mL | |
| CD3ζ | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Western Blot | |
| K25-407.69 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse NKG2D protein | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.