Learn More
CD26 Mouse anti-Human, PE, Clone: L272, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 340423
Product Code. 15835878
  • € 448.00 / Each
Description
CD26, clone L272, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with E1αPGF/JY cells.
CD26 recognizes a 120-kdalton (kDa) antigen that is identical to the enzyme dipeptidyl peptidase (DPP), a serine protease. It is associated with the binding of the TAT transactivating protein of the human immunodeficiency virus (HIV). CD26 and CD45 acts in a co-stimulatory fashion on T lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD26
Monoclonal
PE
Mouse
50 Tests
Immunology
Human
IgG2a κ
Flow Cytometry
L272
Phosphate buffered saline with gelatin and 0.1% sodium azide.
E1aPGF/JY cells
RUO
Primary
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only.