Learn More
CD27 Mouse anti-Human, Alexa Fluor 700, Clone: M-T271, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560611
Description
The M-T271 monoclonal antibody specifically binds to CD27. CD27 presents asa type I transmembrane, disulphide-linked 110 kDa homodimer comprised of two polypeptide chains. The CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on a subset of human thymocytes and on the majority of mature T lymphocytes, activated B cells and NK cells. CD27 is highly induced on T cells after TCR stimulation. CD27 binds to CD70 (also known as, CD27 ligand or CD27L) and may be involved in cellular interaction of T and B lymphocytes.
Specifications
| CD27 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| M-T271 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Human T-CLL cells | |
| 50 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.