missing translation for 'onlineSavingsMsg'
Learn More

CD274 Mouse anti-Human, Brilliant Violet 650, Clone: MIH1, BD

Product Code. 15853049
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15853049 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15853049 Supplier BD Biosciences Supplier No. 563740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PD-L1 and PD-L2 are newly discovered members of the B7 family are the ligands for the program death 1 (PD-1) receptor. They are expressed on immature dendritic cells and mature dendritic cells. PD-L1 is expressed on antigen-presenting cells, including monocytes stimulated with IFN- γ, and activated human and murine dendritic cells. Monoclonal antibodies that block PD-L1 and PD-L2 on dendritic cells result in enhance T cell proliferation and cytokine production. PD-L1 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. Studies show overlapping functions of PD-L1 and PD-L2 and indicate an important role for the PD-L:PD-1 pathway in regulating T cell responses.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274
Applications Flow Cytometry
Classification Monoclonal
Clone MIH1
Conjugate Brilliant Violet 650
Description B7-H1, PD-L1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD274 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.