missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD279 (PD-1), BV711; Clone: EH12.1; BD
Brand: BD Biosciences 564017
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: EH12.1
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human PD-1 Recombinant Protein
Flow cytometry
Specifications
| CD279 (PD-1) | |
| EH12.1 | |
| Human PD-1 Recombinant Protein | |
| 50 Tests | |
| Primary |
| Monoclonal | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction