missing translation for 'onlineSavingsMsg'
Learn More

anti-CD28, Clone: 2C1, Abnova™

Product Code. 16033745
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16033745 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16033745 Supplier Abnova Supplier No. H00000940M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD28
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CD28.
Formulation PBS with no preservative; pH 7.4
Gene CD28
Gene Accession No. NM_006139.3
Gene Alias MGC138290/Tp44
Gene Symbols CD28
Host Species Mouse
Immunogen CD28 (NP_006130.1, 18 a.a. ∼ 152 a.a) partial recombinant protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.