Learn More
CD28 Mouse anti-Human, Unlabeled, Clone: L293, BD
Description
The CD28 antibody, clone L293, is derived from hybridization of Sp2/0-Ag14 mouse myeloma cells with spleen cells from BALB/c mice immunized with the HPB-ALL T-cell line.
The CD28 antigen, a disulfide-linked homodimeric glycoprotein, Mr 44 kilodaltons (kd), is a cell-adhesion molecule (CAM) and functions as the ligand for CD80 (B7-1) and CD86 (B7-2) antigens, which are present on activated B lymphocytes, monocytes, and dendritic cells. Interaction of the CD28 antigen with CD80 or CD86 antigens, or both, co-stimulates CD2 and CD3 antigen/T-cell antigen receptor (TCR)–dependent T-cell–mediated proliferation and cytotoxicity.
Specifications
Specifications
| Antigen | CD28 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | L293 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 200 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.