missing translation for 'onlineSavingsMsg'
Learn More

CD29 Mouse anti-Human, Unlabeled, Clone: MAR4, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555442

Product Code. 15878168

  • ONLINE EXCLUSIVE PRICE
    € 121.00
    € 118.13 / Each
    Save € 2.87
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MAR4 monoclonal antibody specifically binds to CD29. CD29 is a 130 kDa integrin β1 subunit that is expressed as a heterodimeric complex with one of six distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The β1 subunit has a broad tissue distribution; it is expressed on lymphocytes, monocytes and weakly on granulocytes, but not on erythrocytes. These receptors are involved in a variety of cell-cell and cell-matrix interactions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD29
Monoclonal
0.5mg/mL
ITGB1; Integrin beta-1; Integrin β1; FNRB; GPIIA; ITB1; MDF2; MSK12; VLA-4β
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
Mar4
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Purified Human α5β1 Fibronectin Receptor
0.1 mg
Immunology
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.