Learn More
CD3 Mouse anti-Human, Alexa Fluor 700, Clone: SP34-2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557917
Description
Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cells markers. SP34-2 is also capable of inducing cell proliferation on both human and non-human primate PBMC.
Specifications
| CD3 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Purified Human CD3ε Protein | |
| 0.1 mg | |
| Immunology | |
| Baboon, Cynomolgus, Rhesus Monkey | |
| IgG1 λ |
| Flow Cytometry | |
| SP34-2 | |
| Alexa Fluor 700 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.