missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3d Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528313
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat membrane. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD3d Polyclonal antibody specifically detects CD3d in Human samples. It is validated for Western Blot.Specifications
CD3d | |
Polyclonal | |
Unconjugated | |
Cd3d | |
CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 delta; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; IMD19; OKT3, delta chain; T3D; T-cell receptor T3 delta chain; T-cell surface glycoprotein CD3 delta chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P04234 | |
Cd3d | |
Recombinant fragment corresponding to a region within amino acids 1 and 171 of Human CD3 delta | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |