Learn More
CD3 Molecular Complex Rat anti-Mouse, FITC, Clone: 17A2, BD
Description
The 17A2 monoclonal antibody specifically binds to the T-cell receptor-associated CD3 complex that is expressed on many thymocytes and mature T lymphocytes. Plate-bound 17A2 antibody has been reported to induce IL-2 production by cultured T cells in the absence of accessory cells. The binding of the 17A2 antibody to T cells can be blocked by the anti-CD3e mAb 145-2C11. This suggests that the 17A2 antibody recognizes an epitope of the CD3 epsilon chain. In vivo treatment with 17A2 antibody has been reported to partially deplete T lymphocytes and temporarily down-modulates CD3 expression on T cells.
Specifications
Specifications
| Antigen | CD3 Molecular Complex |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 17A2 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Formulation | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Host Species | Rat |
| Immunogen | γδ TCR-positive T-T hybridoma D1 |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.