Learn More
CD3 Mouse anti-Human, V450, Clone: SP34-2, BD
Description
Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cells markers. SP34-2 is also capable of inducing cell proliferation on both human and non-human primate PBMC. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: SP34-2
Isotype: IgG1 λ
Species Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | CD3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SP34-2 |
| Conjugate | Violet Fluor 450 (V450) |
| Formulation | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Purified Human CD3ε Protein |
| Purification Method | Affinity Purified |
| Quantity | 120 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.