Learn More
CD31 Mouse anti-Rat, PE, Clone: TLD-3A12, BD
Description
The TLD-3A12 antibody reacts with CD31, also known as PECAM-1 (platelet endothelial cell adhesion molecule). CD31 is a ∽130 kDa integral membrane glycoprotein, a member of the immunoglobulin super family, that mediates homophilic and heterophilic cell-cell adhesion. CD31 is expressed on endothelial cells, platelets, and subsets of leukocytes. In the human and mouse, multiple alternatively spliced isoforms have been identified, and this alternative splicing may be involved in the regulation of ligand specificity. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis in the mouse and rat. In addition, PECAM-1 has been demonstrated to play an important role in extravasation of leukocytes in vivo, but in vivo treatment with TLD-3A12 mAb had no discernable effect upon the inflammatory infiltrate in experimental allergic encephalomyelitis. The TLD-3A12 mAb partially blocks the proliferation response of antigen-specific CD4+ T cells to antigen-presenting cells and relevant antigen. This mouse anti-rat CD31 antibody cross-reacts with pig endothelial cells and platelets, as determined by immunohistochemical staining of acetone-fixed frozen sections and immunofluorescent staining with flow cytometric analysis, respectively. A suspension of Lewis rat microglial cells were used as the source of the immunogen.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Specifications
| Antigen | CD31 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TLD-3A12 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Description | PECAM-1 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Lewis Rat Microglia |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.