Learn More
CD314 Rat anti-Mouse, APC, Clone: CX5, BD
Description
The CX5 monoclonal antibody specifically binds to NKG2D, also known as CD314. NKG2D is a lectin-like receptor that is detected on resting and IL-2-activated NK cells, activated CD8-positive T lymphocytes, and LPS-activated macrophages, but not on resting T cells or unstimulated macrophages. NKG2D has little homology to the other members of the NKG2 family, NKG2A, C, and E, and does not form heterodimers with CD94. On NK cells, NKG2D is an activating receptor that associates with DAP10, an adapter protein that stimulates the PI3 kinase pathway. An isoform of mouse NKG2D can also associate with the signaling adapter protein DAP12, also known as KARAP ( K iller cell- A ssociated R eceptor- olypeptide), that activates the Syk and ZAP70 tyrosine kinases. On cytotoxic T cells, NKG2D is a co-stimulatory receptor that associates with DAP10. The ligands for NKG2D include the minor histocompatibility antigen H60, MULT1 ( M urine UL 16-binding protein- L ike T ranscript ), and the five retinoic acid-inducible proteins Rae-1 a , b , g , d and e . Interactions of NKG2D with its ligands are involved in the regulation of innate and immune cytotoxic responses to tumor and pathogen-infected cells and in diabetes progression in the NOD mouse. The CX5 mAb blocks the binding of NKG2D to its ligands.
Specifications
Specifications
| Antigen | CD314 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CX5 |
| Concentration | 0.2mg/mL |
| Conjugate | APC |
| Description | Klrk1; NKG2D; NKG2-D; NK cell receptor D; Natural killer cell group 2D |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Rat |
| Immunogen | Purified Mouse NKG2D protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.