missing translation for 'onlineSavingsMsg'
Learn More

CD4 v4 Mouse anti-Human, PE, Clone: L120, BD

Product Code. 15805878
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15805878 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805878 Supplier BD Biosciences Supplier No. 340419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD4 v4, clone L120, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with human peripheral blood lymphocytes.

CD4 recognizes an antigen, approximately 59 kdaltons (kDa), that is present on the helper-inducer subset of T lymphocytes and monocytes. The CD4 antigen is involved in the class II recognition of foreign antigens and has been identified as a receptor for human immunodeficiency virus (HIV). Clone L120 recognizes amino acids 306-370 of the antigen and does not block the binding of the gp120 of HIV, but inhibits the proliferation of CD4 T lymphocytes to class II antigens.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4v4
Applications Flow Cytometry
Classification Monoclonal
Clone L120
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human peripheral blood lymphocytes
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.