Learn More
CD42a Mouse anti-Human, PerCP, Clone: BEB1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340537
Description
The CD42a antibody, clone Beb1, is derived from hybridization of Sp2/0 mouse myeloma cells with spleen cells from BALB/c mice immunized with normal human platelets.
The CD42a antibody recognizes a 17 to 22-kilodalton (kDa) single-chain, platelet membrane glycoprotein, also known as gpIX that forms a noncovalent complex with gpIb and gpV. Glycoprotein Ib consists of an α–chain disulfide-linked to a β chain. Glycoproteins Ibα (140 to 145 kDa), Ibβ (24 to 25 kDa), and V (82 to 86 kDa) have been designated as CD42b, CD42c and CD42d respectively. CD42a has been reported to react with both gpIX and the gpIb-IX-V complex.
Specifications
| CD42a | |
| Monoclonal | |
| 12.5μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human platelets | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| BEB1 | |
| PerCP | |
| Mouse | |
| 50 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.