Learn More
CD45 Mouse anti-Human, Brilliant Violet 711, Clone: HI30, BD
Mouse Monoclonal Antibody
€ 203.30 - € 402.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | HI30 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 711 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15873569
|
BD Biosciences
564358 |
25 Tests |
€ 209.00 € 203.30 / Each Save € 5.70 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15863569
|
BD Biosciences
564357 |
100 Tests |
€ 402.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC ( Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.
Specifications
| CD45 | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| IgG1 κ | |
| Affinity Purified | |
| PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5 |
| HI30 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Peripheral Blood Leucocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.