missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, PE-CF594, Clone: HI30, BD

Mouse Monoclonal Antibody

€ 132.68 - € 474.00

Specifications

Antigen CD45
Clone HI30
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-CF594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15841809
View Documents
BD Biosciences
562312
25 Tests
ONLINE EXCLUSIVE PRICE
€ 133.00
€ 132.68 / Each
Save € 0.32
0.24% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15811779
View Documents
BD Biosciences
562279
100 Tests
ONLINE EXCLUSIVE PRICE
€ 487.00
€ 474.00 / Each
Save € 13.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

Specifications

Specifications

CD45
Flow Cytometry
PE-CF594
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
HI30
Monoclonal
Mouse
Human
Human Peripheral Blood Leucocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.